accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 953039-63-9

IUPAC Name: 8-bromo-2-chloro-6-fluoroquinazoline
Synonyms: 8-bromo-2-chloro-6-fluoroquinazoline
Molecular Weight (g/mol): 261.48
Molecular Formula: C8H3BrClFN2
InChi Key: XTEWVYPXKPYPRE-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C2N=C(Cl)N=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results