accessibility menu, dialog, popup

UserName

Viewing profile as user:
PPPdClClClClFe

CAS RN 95464-05-4

IUPAC Name: Bis(diphenylphosphino)ferrocene]palladium(II) chloride
Synonyms: —
Molecular Weight (g/mol): 816.64
Molecular Formula: C35H30Cl4FeP2Pd
InChi Key: —
SMILES: [Fe].ClCCl.Cl[Pd]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (9)
  • (4)
Quantity 
  • (4)
  • (4)
  • (1)
  • (4)
Physical Form 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (3)
  • (3)
  • (3)
  • (9)
  • (4)
  • (4)
  • (4)
  • (1)
  • (4)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)