accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NHN

CAS RN 956-61-6

IUPAC Name: 1-[(4-tert-butylphenyl)methyl]piperazine
Synonyms: 1-[(4-tert-butylphenyl)methyl]piperazine
Molecular Weight (g/mol): 232.37
Molecular Formula: C15H24N2
InChi Key: UQLCETYSARZZSR-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(CN2CCNCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results