accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONOO

CAS RN 956-75-2

IUPAC Name: 4-nitrophenyl hexanoate
Synonyms: 4-nitrophenyl hexanoate
Molecular Weight (g/mol): 237.26
Molecular Formula: C12H15NO4
InChi Key: OLRXUEYZKCCEKK-UHFFFAOYSA-N
SMILES: CCCCCC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results