accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNOOH

CAS RN 956317-36-5

IUPAC Name: 5-methyl-2-(2H-1,2,3-triazol-2-yl)benzoic acid
Synonyms: 5-methyl-2-(1,2,3-triazol-2-yl)benzoic acid
Molecular Weight (g/mol): 203.2
Molecular Formula: C10H9N3O2
InChi Key: SRBAGFIYKNQXDV-UHFFFAOYSA-N
SMILES: CC1=CC(C(O)=O)=C(C=C1)N1N=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results