accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CO

CAS RN 957-51-7

IUPAC Name: N,N-dimethyl-2,2-diphenylacetamide
Synonyms: diphenamid
Molecular Weight (g/mol): 239.32
Molecular Formula: C16H17NO
InChi Key: QAHFOPIILNICLA-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results