accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 957-68-6

IUPAC Name: (6R,7R)-3-[(acetyloxy)methyl]-7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Synonyms: 7β-aminocephalosporanic acid 7-aca 7-aminocephalosporinic acid 7-aminocephalosporanic acid
Molecular Weight (g/mol): 272.28
Molecular Formula: C10H12N2O5S
InChi Key: HSHGZXNAXBPPDL-HZGVNTEJSA-N
SMILES: CC(=O)OCC1=C(N2[C@H](SC1)[C@H](N)C2=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (4)