accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 957121-05-0

IUPAC Name: (3-cyano-2-fluorophenyl)boronic acid
Synonyms: 3-cyano-2-fluorophenylboronic acid
Molecular Weight (g/mol): 164.93
Molecular Formula: C7H5BFNO2
InChi Key: HENIWPFEWBREIB-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C(=CC=C1)C#N
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results