accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95715-87-0

IUPAC Name: tert-butyl (4R)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
Synonyms: tert-butyl (4R)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
Molecular Weight (g/mol): 229.28
Molecular Formula: C11H19NO4
InChi Key: PNJXYVJNOCLJLJ-QMMMGPOBSA-N
SMILES: CC(C)(C)OC(=O)N1[C@H](COC1(C)C)C=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (3)
Boiling Point 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)