accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNBrCl

CAS RN 957187-27-8

IUPAC Name: 8-bromo-6-chloroimidazo[1,2-a]pyridine
Synonyms: 8-bromo-6-chloroimidazo[1,2-a]pyridine
Molecular Weight (g/mol): 231.48
Molecular Formula: C7H4BrClN2
InChi Key: LMPPBTNPACXNDI-UHFFFAOYSA-N
SMILES: ClC1=CN2C=CN=C2C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results