accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95798-23-5

IUPAC Name: benzyl 4-hydroxypiperidine-1-carboxylate
Synonyms: benzyl 4-hydroxypiperidine-1-carboxylate
Molecular Weight (g/mol): 235.28
Molecular Formula: C13H17NO3
InChi Key: JKIUUDJOCYHIGY-UHFFFAOYSA-N
SMILES: OC1CCN(CC1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results