accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95838-16-7

IUPAC Name: λ⁴-cerium(4+) disulfate
Synonyms: cerium(IV) sulfate
Molecular Weight (g/mol): 332.23
Molecular Formula: CeO8S2
InChi Key: VZDYWEUILIUIDF-UHFFFAOYSA-J
SMILES: [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)