accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 958852-01-2

IUPAC Name: 5-{[4-(pyridin-4-yl)quinolin-6-yl]methylidene}-1,3-thiazolidine-2,4-dione
Synonyms: 5-{[4-(pyridin-4-yl)quinolin-6-yl]methylidene}-1,3-thiazolidine-2,4-dione
Molecular Weight (g/mol): 333.37
Molecular Formula: C18H11N3O2S
InChi Key: QDITZBLZQQZVEE-UHFFFAOYSA-N
SMILES: O=C1NC(=O)C(S1)=CC1=CC=C2N=CC=C(C3=CC=NC=C3)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results