accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 959-23-9

IUPAC Name: (2E)-1-(4-methoxyphenyl)-3-phenylprop-2-en-1-one
Synonyms: 4'-methoxychalcone
Molecular Weight (g/mol): 238.29
Molecular Formula: C16H14O2
InChi Key: KJHHAPASNNVTSN-KPKJPENVSA-N
SMILES: COC1=CC=C(C=C1)C(=O)\C=C\C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results