accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 959-26-2

IUPAC Name: 1,4-bis(2-hydroxyethyl) benzene-1,4-dicarboxylate
Synonyms: 1,4-bis(2-hydroxyethyl) benzene-1,4-dicarboxylate
Molecular Weight (g/mol): 254.24
Molecular Formula: C12H14O6
InChi Key: QPKOBORKPHRBPS-UHFFFAOYSA-N
SMILES: OCCOC(=O)C1=CC=C(C=C1)C(=O)OCCO
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results