accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 959-52-4

IUPAC Name: 1-[3,5-bis(prop-2-enoyl)-1,3,5-triazinan-1-yl]prop-2-en-1-one
Synonyms: 1-[3,5-bis(prop-2-enoyl)-1,3,5-triazinan-1-yl]prop-2-en-1-one
Molecular Weight (g/mol): 249.27
Molecular Formula: C12H15N3O3
InChi Key: FYBFGAFWCBMEDG-UHFFFAOYSA-N
SMILES: C=CC(=O)N1CN(CN(C1)C(=O)C=C)C(=O)C=C
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results