accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3COOCH3

CAS RN 959303-25-4

IUPAC Name: N-ethyl-4-methoxy-N-(3-methylphenyl)benzamide
Synonyms: N-ethyl-4-methoxy-N-(3-methylphenyl)benzamide
Molecular Weight (g/mol): 269.34
Molecular Formula: C17H19NO2
InChi Key: DFPGANUPVBKLNA-UHFFFAOYSA-N
SMILES: CCN(C(=O)C1=CC=C(OC)C=C1)C1=CC=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results