accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 96-05-9

IUPAC Name: prop-2-en-1-yl 2-methylprop-2-enoate
Synonyms: allyl methacrylate
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: FBCQUCJYYPMKRO-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCC=C
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)