accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHBrBr

CAS RN 96-13-9

IUPAC Name: 2,3-dibromopropan-1-ol
Synonyms: 2,3-dibromo-1-propanol
Molecular Weight (g/mol): 217.89
Molecular Formula: C3H6Br2O
InChi Key: QWVCIORZLNBIIC-UHFFFAOYNA-N
SMILES: OCC(Br)CBr
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)