CAS RN 96-32-2
Methyl bromoacetate, 98+%
CAS: 96-32-2 Molecular Formula: C3H5BrO2 Molecular Weight (g/mol): 152.975 MDL Number: MFCD00000189 InChI Key: YDCHPLOFQATIDS-UHFFFAOYSA-N Synonym: methyl bromoacetate,bromoacetic acid methyl ester,methyl monobromoacetate,acetic acid, bromo-, methyl ester,methyl alpha-bromoacetate,methylbromoacetate,acetic acid, 2-bromo-, methyl ester,methylester kyseliny bromoctove,methyl .alpha.-bromoacetate,methylester kyseliny bromoctove czech PubChem CID: 60984 IUPAC Name: methyl 2-bromoacetate SMILES: COC(=O)CBr
Methyl bromoacetate, 99%
CAS: 96-32-2 Molecular Formula: C3H5BrO2 Molecular Weight (g/mol): 152.98 MDL Number: MFCD00000189 InChI Key: YDCHPLOFQATIDS-UHFFFAOYSA-N Synonym: methyl bromoacetate,bromoacetic acid methyl ester,methyl monobromoacetate,acetic acid, bromo-, methyl ester,methyl alpha-bromoacetate,methylbromoacetate,acetic acid, 2-bromo-, methyl ester,methylester kyseliny bromoctove,methyl .alpha.-bromoacetate,methylester kyseliny bromoctove czech PubChem CID: 60984 IUPAC Name: methyl 2-bromoacetate SMILES: COC(=O)CBr
Methyl Bromoacetate 98.0+%, TCI America™
CAS: 96-32-2 Molecular Formula: C3H5BrO2 Molecular Weight (g/mol): 152.975 MDL Number: MFCD00000189 InChI Key: YDCHPLOFQATIDS-UHFFFAOYSA-N Synonym: methyl bromoacetate,bromoacetic acid methyl ester,methyl monobromoacetate,acetic acid, bromo-, methyl ester,methyl alpha-bromoacetate,methylbromoacetate,acetic acid, 2-bromo-, methyl ester,methylester kyseliny bromoctove,methyl .alpha.-bromoacetate,methylester kyseliny bromoctove czech PubChem CID: 60984 IUPAC Name: methyl 2-bromoacetate SMILES: COC(=O)CBr
Sigma Aldrich Methyl bromoacetate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc Methyl Bromoacetate, MP Biomedicals
CAS: 96-32-2 Molecular Formula: C3H5BrO2 Molecular Weight (g/mol): 152.975 InChI Key: YDCHPLOFQATIDS-UHFFFAOYSA-N Synonym: methyl bromoacetate,bromoacetic acid methyl ester,methyl monobromoacetate,acetic acid, bromo-, methyl ester,methyl alpha-bromoacetate,methylbromoacetate,acetic acid, 2-bromo-, methyl ester,methylester kyseliny bromoctove,methyl .alpha.-bromoacetate,methylester kyseliny bromoctove czech PubChem CID: 60984 IUPAC Name: methyl 2-bromoacetate SMILES: COC(=O)CBr