accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCl

CAS RN 96-34-4

IUPAC Name: methyl 2-chloroacetate
Synonyms: methyl chloroacetate
Molecular Weight (g/mol): 108.52
Molecular Formula: C3H5ClO2
InChi Key: QABLOFMHHSOFRJ-UHFFFAOYSA-N
SMILES: COC(=O)CCl
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)