accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HOSHOOONOO

CAS RN 96-67-3

IUPAC Name: 3-amino-2-hydroxy-5-nitrobenzene-1-sulfonic acid
Synonyms: 3-amino-2-hydroxy-5-nitrobenzenesulfonic acid
Molecular Weight (g/mol): 234.18
Molecular Formula: C6H6N2O6S
InChi Key: DQIVFTJHYKDOMZ-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(=C1O)S(O)(=O)=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results