accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 96-96-8

IUPAC Name: 4-methoxy-2-nitroaniline
Synonyms: fast bordeaux GP
Molecular Weight (g/mol): 168.15
Molecular Formula: C7H8N2O3
InChi Key: QFMJFXFXQAFGBO-UHFFFAOYSA-N
SMILES: COC1=CC=C(N)C(=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (2)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)