accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 960-71-4

IUPAC Name: triphenylborane
Synonyms: triphenylborane
Molecular Weight (g/mol): 242.13
Molecular Formula: C18H15B
InChi Key: MXSVLWZRHLXFKH-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)B(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)