CAS RN 96017-59-3
IUPAC Name:
2-(prop-2-yn-1-yl)octanoic acid
Synonyms:
2-(prop-2-yn-1-yl)octanoic acid
Molecular Weight (g/mol):
182.26
Molecular Formula:
C11H18O2
InChi Key:
DUQSBRQHALCSLC-UHFFFAOYNA-N
SMILES:
CCCCCCC(CC#C)C(O)=O
1
–
1
of
1
results
1
2-Hexyl-4-pentynoic Acid 95.0+%, TCI America™
CAS: 96017-59-3 Molecular Formula: C11H18O2 Molecular Weight (g/mol): 182.26 MDL Number: MFCD00946863 InChI Key: DUQSBRQHALCSLC-UHFFFAOYNA-N Synonym: 2-Propargyloctanoic Acid, 2-(2-Propynyl)octanoic Acid PubChem CID: 175664 IUPAC Name: 2-(prop-2-yn-1-yl)octanoic acid SMILES: CCCCCCC(CC#C)C(O)=O