CAS RN 96099-84-2
CAS RN 96099-84-2
IUPAC Name:
methyl 2-{[(tert-butoxy)carbonyl]amino}-3-hydroxybutanoate
Synonyms:
methyl 2-[(tert-butoxycarbonyl)amino]-3-hydroxybutanoate
Molecular Weight (g/mol):
233.26
Molecular Formula:
C10H19NO5
InChi Key:
MZMWAPNVRMDIPS-UHFFFAOYNA-N
SMILES:
COC(=O)C(NC(=O)OC(C)(C)C)C(C)O
1
–
1
of
1
results
1
N-(tert-Butoxycarbonyl)-D-threonine Methyl Ester 98.0+%, TCI America™
CAS: 96099-84-2 Molecular Formula: C10H19NO5 Molecular Weight (g/mol): 233.264 InChI Key: MZMWAPNVRMDIPS-NKWVEPMBSA-N Synonym: N-Boc-D-threonine Methyl Ester, Boc-D-Thr-OMe PubChem CID: 40501164 IUPAC Name: methyl (2R,3S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoate SMILES: CC(C(C(=O)OC)NC(=O)OC(C)(C)C)O