accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NH2H3CH3CCH3CH3H3CCH3

CAS RN 961-38-6

IUPAC Name: 2,4,6-tri-tert-butylaniline
Synonyms: 2,4,6-tri-tert-butylaniline
Molecular Weight (g/mol): 261.45
Molecular Formula: C18H31N
InChi Key: REJGDSCBQPJPQT-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=C(N)C(=C1)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)