CAS RN 96100-94-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
[5]Cycloparaphenylene 95.0+%, TCI America™
CAS: 96100-94-6 Molecular Formula: C30H20 Molecular Weight (g/mol): 380.49 InChI Key: JPDIIJOQNLNRQS-UHFFFAOYSA-N Synonym: [5]CPP PubChem CID: 131637028 SMILES: C1CC2=C=C=C1C3=C=C=C(CC3)C4=C=C=C(CC4)C5=CC=C(C=C5)C6=C=C=C2CC6