accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 96107-95-8

IUPAC Name: heptacyclo[31.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹]dotetraconta-1(37),3,5,7(42),9,11,13(41),15,17,19(40),21,23,25(39),27,29,31(38),33,35-octadecaene-37,38,39,40,41,42-hexol
Synonyms: heptacyclo[31.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹]dotetraconta-1(37),3,5,7(42),9,11,13(41),15,17,19(40),21,23,25(39),27,29,31(38),33,35-octadecaene-37,38,39,40,41,42-hexol
Molecular Weight (g/mol): 636.74
Molecular Formula: C42H36O6
InChi Key: JLSWUKWFQCVKCL-UHFFFAOYSA-N
SMILES: OC1=C2CC3=CC=CC(CC4=CC=CC(CC5=CC=CC(CC6=CC=CC(CC7=CC=CC(CC1=CC=C2)=C7O)=C6O)=C5O)=C4O)=C3O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)