CAS RN 96314-98-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
FURA 2 pentasodium salt
CAS: 96314-98-6 Molecular Weight (g/mol): 751.45 MDL Number: MFCD00083328