CAS RN 96404-79-4
CAS RN 96404-79-4
IUPAC Name:
triphenylene-2-carbaldehyde
Synonyms:
triphenylene-2-carbaldehyde
Molecular Weight (g/mol):
256.3
Molecular Formula:
C19H12O
InChi Key:
NXDGVEYXRSKONY-UHFFFAOYSA-N
SMILES:
O=CC1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1
1
–
1
of
1
results
1
2-Triphenylenecarboxaldehyde 98.0+%, TCI America™
CAS: 96404-79-4 Molecular Formula: C19H12O Molecular Weight (g/mol): 256.30 MDL Number: MFCD29089374 InChI Key: NXDGVEYXRSKONY-UHFFFAOYSA-N PubChem CID: 13375435 IUPAC Name: triphenylene-2-carbaldehyde SMILES: O=CC1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1