accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNCH3HOH3CHOCH3OHOHTi

CAS RN 96471-81-7

IUPAC Name: 1-({2-[bis(2-hydroxypropyl)amino]ethyl}(2-hydroxypropyl)amino)propan-2-ol titanium
Synonyms: quadrol titanium
Molecular Weight (g/mol): 340.29
Molecular Formula: C14H32N2O4Ti
InChi Key: ILOYOQJSNBNOPE-UHFFFAOYNA-N
SMILES: [Ti].CC(O)CN(CCN(CC(C)O)CC(C)O)CC(C)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results