accessibility menu, dialog, popup

UserName

Viewing profile as user:
FClO

CAS RN 96515-79-6

IUPAC Name: 5-chloro-2-fluorobenzaldehyde
Synonyms: 5-chloro-2-fluorobenzaldehyde
Molecular Weight (g/mol): 158.56
Molecular Formula: C7H4ClFO
InChi Key: WDTUCEMLUHTMCB-UHFFFAOYSA-N
SMILES: FC1=CC=C(Cl)C=C1C=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)