accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 96797-15-8

IUPAC Name: 4-iodo-1-(triphenylmethyl)-1H-imidazole
Synonyms: 4-iodo-1-(triphenylmethyl)imidazole
Molecular Weight (g/mol): 436.3
Molecular Formula: C22H17IN2
InChi Key: DXJZJYPLPZEYBH-UHFFFAOYSA-N
SMILES: IC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)