accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNNCH3

CAS RN 96897-04-0

IUPAC Name: 4-methyl-4'-(2-{4'-methyl-[2,2'-bipyridin]-4-yl}ethyl)-2,2'-bipyridine
Synonyms: 4-methyl-4'-(2-{4'-methyl-[2,2'-bipyridin]-4-yl}ethyl)-2,2'-bipyridine
Molecular Weight (g/mol): 366.47
Molecular Formula: C24H22N4
InChi Key: KLTATORHZNMNDJ-UHFFFAOYSA-N
SMILES: CC1=CC(=NC=C1)C1=NC=CC(CCC2=CC(=NC=C2)C2=NC=CC(C)=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results