accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFNH2FFFFFFF(?)

CAS RN 969-25-5

IUPAC Name: nonafluoro-[1,1'-biphenyl]-4-amine
Synonyms: nonafluoro-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 331.14
Molecular Formula: C12H2F9N
InChi Key: DVKUPHGYOMHVDR-UHFFFAOYSA-N
SMILES: NC1=C(F)C(F)=C(C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results