accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHOSOHOO

CAS RN 97-05-2

IUPAC Name: 2-hydroxy-5-sulfobenzoic acid
Synonyms: sulfosalicylic acid 2-hydroxysulfo-benzoic acid kalcolor anodizing acid phlogosam phlogosol benzoic acid, 2-hydroxysulfo- beryllium salt (1:1) sulfosalicylic acid sulphosalicylic acid salicylic acid, sulfo- cesium 5-sulfosalicylate
Molecular Weight (g/mol): 218.18
Molecular Formula: C7H6O6S
InChi Key: YCPXWRQRBFJBPZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Grade 
  • (2)
pH 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)