accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPSOCH3OClCl

CAS RN 97-17-6

IUPAC Name: O-2,4-dichlorophenyl O,O-diethyl phosphorothioate
Synonyms: dichlofenthion
Molecular Weight (g/mol): 315.14
Molecular Formula: C10H13Cl2O3PS
InChi Key: WGOWCPGHOCIHBW-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)OC1=CC=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results