accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 97-24-5

IUPAC Name: 4-chloro-2-[(5-chloro-2-hydroxyphenyl)sulfanyl]phenol
Synonyms: novex caswell no. 851 fenticlor ovitrol meflorin fenticloro fenticlorum S 7 (antimycotic) epa pesticide chemical code 064209 C12H8cl2O2S
Molecular Weight (g/mol): 287.15
Molecular Formula: C12H8Cl2O2S
InChi Key: ANUSOIHIIPAHJV-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1SC1=CC(Cl)=CC=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results