accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 97145-88-5

H3CHNClHCl

CAS RN 97145-88-5

IUPAC Name: (3-chloropropyl)(methyl)amine hydrochloride
Synonyms: (3-chloropropyl)(methyl)amine hydrochloride
Molecular Weight (g/mol): 144.04
Molecular Formula: C4H11Cl2N
InChi Key: SHBWHBGVZIDIQY-UHFFFAOYSA-N
SMILES: Cl.CNCCCCl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results