accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOBr

CAS RN 97511-06-3

IUPAC Name: 1-(1-benzothiophen-2-yl)-2-bromoethan-1-one
Synonyms: 1-(1-benzothiophen-2-yl)-2-bromoethanone
Molecular Weight (g/mol): 255.13
Molecular Formula: C10H7BrOS
InChi Key: WVTSOGFICBVCRE-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results