CAS RN 97534-21-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Merbarone
A cell-permeable anticancer drug that inhibits the catalytic activity of topoisomerase II (topo II) without damaging DNA or stabilizing DNA-topo II cleavable complexes.