CAS RN 978-65-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-[3,5-Bis(trifluoromethyl)phenyl]-3-(4-fluorophenyl)urea, 97%, Thermo Scientific™
CAS: 978-65-4 MDL Number: MFCD04069619