CAS RN 97892-67-6
CAS RN 97892-67-6
IUPAC Name:
2-chloro-7-methoxy-4-methylquinoline
Synonyms:
2-chloro-7-methoxy-4-methylquinoline
Molecular Weight (g/mol):
207.66
Molecular Formula:
C11H10ClNO
InChi Key:
SMJPQHMCVFWKMI-UHFFFAOYSA-N
SMILES:
COC1=CC=C2C(C)=CC(Cl)=NC2=C1
1
–
1
of
1
results
1
2-Chloro-7-methoxy-4-methylquinoline, 97%, Thermo Scientific Chemicals
CAS: 97892-67-6 Molecular Formula: C11H10ClNO Molecular Weight (g/mol): 207.657 MDL Number: MFCD00711757 InChI Key: SMJPQHMCVFWKMI-UHFFFAOYSA-N Synonym: 2-chloro-7-methoxy-4-methyl-quinoline,enamine_005386,2-chloranyl-7-methoxy-4-methyl-quinoline PubChem CID: 732214 IUPAC Name: 2-chloro-7-methoxy-4-methylquinoline SMILES: CC1=CC(=NC2=C1C=CC(=C2)OC)Cl