CAS RN 97925-43-4
IUPAC Name:
4-bromo-1,2-oxazole
Synonyms:
4-bromo-1,2-oxazole
Molecular Weight (g/mol):
147.96
Molecular Formula:
C3H2BrNO
InChi Key:
IRDVIGFQMLUJAM-UHFFFAOYSA-N
SMILES:
BrC1=CON=C1
1
–
2
of
2
results
1
Promotions Available
4-Bromoisoxazole, 95%
4-Bromoisoxazole, 95%
CAS: 97925-43-4 Molecular Formula: C3H2BrNO Molecular Weight (g/mol): 147.96 InChI Key: IRDVIGFQMLUJAM-UHFFFAOYSA-N Synonym: 4-bromoisoxazole,4-bormoisoxazole,isoxazole, 4-bromo,isoxazole, 4-bromo-6ci,9ci,4-bromo isoxazole,4-bromo-isoxazole,zlchem 1329 PubChem CID: 3862248 IUPAC Name: 4-bromo-1,2-oxazole SMILES: C1=C(C=NO1)Br
2
Sigma Aldrich 4-Bromoisoxazole
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.