accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO

CAS RN 98-00-0

IUPAC Name: (furan-2-yl)methanol
Synonyms: furfuryl alcohol 2-furane-methanol furfuryl alkohol furylcarbinol furfural alcohol 2-furfuryl alcohol furfuranol
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: XPFVYQJUAUNWIW-UHFFFAOYSA-N
SMILES: OCC1=CC=CO1
Molecular Weight (g/mol) 
  • (7)
  • (6)
Boiling Point 
  • (6)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (10)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (1)
  • (2)
  • (7)
  • (7)
  • (6)
  • (6)
  • (3)
  • (3)
  • (2)
  • (10)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (9)
  • (1)