accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 98-01-1

IUPAC Name: furan-2-carbaldehyde
Synonyms: bran oil caswell no. 466 furole furfurole furfurale artificial oil of ants 2-furfuraldehyde furale epa pesticide chemical code 043301 2-furfural
Molecular Weight (g/mol): 96.09
Molecular Formula: C5H4O2
InChi Key: HYBBIBNJHNGZAN-UHFFFAOYSA-N
SMILES: O=CC1=CC=CO1
Molecular Weight (g/mol) 
  • (6)
  • (12)
Boiling Point 
  • (4)
  • (2)
  • (1)
  • (10)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
Physical Form 
  • (8)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (8)
  • (1)
  • (6)
  • (12)
  • (4)
  • (2)
  • (1)
  • (10)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (8)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)