accessibility menu, dialog, popup

UserName

Viewing profile as user:
HSO

CAS RN 98-02-2

IUPAC Name: (furan-2-yl)methanethiol
Synonyms: 2-furanmethanethiol
Molecular Weight (g/mol): 114.16
Molecular Formula: C5H6OS
InChi Key: ZFFTZDQKIXPDAF-UHFFFAOYSA-N
SMILES: SCC1=CC=CO1
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (1)
Boiling Point 
  • (4)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)