accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3

CAS RN 98-06-6

IUPAC Name: tert-butylbenzene
Synonyms: T-butylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: YTZKOQUCBOVLHL-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (5)
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (1)
  • (2)
  • (5)
  • (6)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (8)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)
  • (1)