accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFF

CAS RN 98-08-8

IUPAC Name: (trifluoromethyl)benzene
Synonyms: (trifluoromethyl)-benzene CF3ph usaf MA-16 α,α,α-trifluorotoluene phenylfluoroform PhCF3 ω-trifluorotoluene
Molecular Weight (g/mol): 146.11
Molecular Formula: C7H5F3
InChi Key: GETTZEONDQJALK-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Boiling Point 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (3)
  • (6)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)